missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-LKB1 (Ser428) Polyclonal Antibody
Phospho-LKB1 (Ser428) Polyclonal Antibody
Brand: Invitrogen PA536858
This item is not returnable.
View return policy
Description
Phospho-LKB1 (Ser428) Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| Phospho-LKB1 (Ser428) | |
| Polyclonal | |
| Unconjugated | |
| STK11 | |
| AA408040; CAMK group; CAMKL family; hLKB1; liver kinase B1; Liver kinase B1 homolog; LKB 1; LKB1; LKB-1; LKB1(S); Par-4; PJS; polarization-related protein LKB1; R75140; renal carcinoma antigen NY-REN-19; serine/threonine kinase 11; serine/threonine-protein kinase 11; serine/threonine-protein kinase LKB1; serine/threonine-protein kinase STK11; STK11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20869, 314621, 6794 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| D4AE59, Q15831, Q9WTK7 | |
| STK11 | |
| Synthetic phosphopeptide derived from human LKB1 around the phosphorylation site of Serine 428. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction