missing translation for 'onlineSavingsMsg'
Learn More

Phospho-LATS1/LATS2 (Thr1079, Thr1041) Polyclonal Antibody, Invitrogen™

Product Code. 17269723 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17269723 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17269723 Supplier Thermo Scientific Supplier No. PA5117227

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: HeLa

Phospho-LATS1/LATS2 (Thr1079, Thr1041) Polyclonal antibody specifically detects Phospho-LATS1/LATS2 (Thr1079, Thr1041) in Human, Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-LATS1/LATS2 (Thr1079, Thr1041)
Applications Western Blot
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Lats1
Gene Accession No. O95835, Q9NRM7
Gene Alias 4932411G09Rik; AV277261; AW208599; AW228608; h-warts; kinase phosphorylated during mitosis protein; KPM; large tumor suppressor; large tumor suppressor 2; large tumor suppressor homolog 1; Large tumor suppressor homolog 2; large tumor suppressor kinase 1; large tumor suppressor kinase 2; large tumor suppressor, homolog 2; large tumor supressor, homolog 1; LATS (large tumor suppressor, Drosophila) homolog 1; LATS (large tumor suppressor, Drosophila) homolog 2; LATS, large tumor suppressor, homolog 1; LATS, large tumor suppressor, homolog 2; lats1; LATS2; RGD1564085; serine/threonine kinase KPM; Serine/threonine-protein kinase kpm; serine/threonine-protein kinase LATS1; Serine/threonine-protein kinase LATS2; WARTS; WARTS protein kinase; warts-like kinase; wts
Host Species Rabbit
Immunogen A phospho specific peptide corresponding to residues surrounding T1079 of human LATS1
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26524, 9113
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.