missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-LAT (Tyr220) Recombinant Rabbit Monoclonal Antibody (24GB1375)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557159

 View more versions of this product

Product Code. 30275144

  • £434.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Phospho-LAT (Tyr220) Recombinant Monoclonal Antibody for Western Blot

LAT (Linker for Activation of T cells) is a 36-38 kDa double-palmitoylated transmembrane adaptor protein expressed by T cells, pre-B cells, NK cells, mast cells and platelets. After immunoreceptor triggering, LAT becomes multiply tyrosine-phosphorylated by Syk-, Src-, or Tec-family kinases, providing docking sites for downstream signaling molecules. LAT is essential for TCR-dependent T cell- and FcepsilonRI-dependent mast cell activation, as well as for maturation of early thymocytes. It is also involved in NK cell signaling and platelet activation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-LAT (Tyr220)
Recombinant Monoclonal
2.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O43561, O54957, O70601
LAT
A synthesized peptide derived from human Phospho-LAT (Tyr220) (aa 213-227).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot, Immunocytochemistry
24GB1375
Unconjugated
LAT
36 kDa phospho-tyrosine adapter protein; 36 kDa phospho-tyrosine adaptor protein; Lat; LAT1; linker for activation of T cells; linker for activation of T cells, transmembrane adaptor; linker for activation of T-cells; linker for activation of T-cells family member 1; linker protein; p36-38; pp36
Rabbit
Affinity chromatography
RUO
16797, 27040, 81511
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.