missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-KSR2 (Thr497) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105093
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of KSR2 only when phosphorylated at Thr497.
KSR2 serine/threonine kinase or kinase suppressor of Ras 2 specifically regulates the activity of MEKK3 and COT but not other members of the MAP3K family. KSR interacts with a regulatory Raf molecule in cis to induce a conformational switch of MEK, facilitating MEK's phosphorylation by a separate catalytic Raf molecule in trans. Blocks MAP3K8 kinase activity and MAP3K8- mediated signaling. Acts as a negative regulator of MAP3K3- mediated activation of ERK, JNK and NF-kappa-B pathways, inhibiting MAP3K3-mediated interleukin-8 production.
Specifications
| Phospho-KSR2 (Thr497) | |
| Polyclonal | |
| Unconjugated | |
| KSR2 | |
| hKSR2; Kinase suppressor of Ras 2; ksr2; RGD1562674; testes-kinase suppressor of ras 2 isoform | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 283455, 288691, 333050 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3UVC0, Q6VAB6 | |
| KSR2 | |
| A synthesized peptide derived from human KSR2(Accession Q6VAB6), corresponding to amino acid residues around phosphorylated Thr497. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction