missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-KSR1 (Ser569) Polyclonal Antibody

Phospho-KSR1 (Ser569) Polyclonal Antibody
Brand: Invitrogen PA5105675
This item is not returnable.
View return policy
Description
Phospho-KSR1 (Ser569) Polyclonal Antibody for Western Blot
Specifications
| Phospho-KSR1 (Ser569) | |
| Polyclonal | |
| Unconjugated | |
| KSR1 | |
| AW492498; B-KSR1; CAP kinase; D11Bhm183e; D11Bhm184e; hb; HB protein; kinase KSR1; Kinase suppressor of Ras 1; Ksr; KSR1; mKSR1; OTTHUMP00000162931; protein Hb; RSU2 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 16706, 8844 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q61097, Q8IVT5 | |
| KSR1 | |
| A synthesized peptide derived from human KSR1(Accession Q8IVT5), corresponding to amino acid residues around phosphorylated Ser569. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction