Learn More
Phospho-KLF5 (Ser303) Polyclonal Antibody
GREENER_CHOICE
Phospho-KLF5 (Ser303) Polyclonal Antibody
Brand:  Invitrogen PA5105317
Product Code. 16621765
  • £457.00 / 100µL
Description
Phospho-KLF5 (Ser303) Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-KLF5 (Ser303)
Polyclonal
Unconjugated
KLF5
(intestinal Kruppel-like factor; 4930520J07Rik; basic transcription element binding protein 2; basic transcription element binding protein BTEB2; basic transcription element-binding protein 2; BTEB2; BTE-binding protein 2; CKLF; colon krueppel-like factor; colon kruppel-like factor; GC box binding protein 2; GC-box-binding protein 2; IKLF; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; Klf5; Klf5C isoform; Krueppel-like factor 5; Kruppel like factor 5; Kruppel-like factor 5; Kruppel-like factor 5 (intestinal); RP11-505F3.5; transcription factor BTEB2
Rabbit
Sequential Chromatography
RUO
12224, 688, 84410
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q13887, Q9Z0Z7
KLF5
A synthesized peptide derived from human KLF5(Accession Q13887), corresponding to amino acid residues around phosphorylated Ser303.
100 μL
Primary
Human, Mouse, Rat