missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Kir6.2 (Thr224) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540157
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology.
This receptor is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. Can be blocked by extracellular barium. Subunit of ATP-sensitive potassium channels (KATP). Can form cardiac and smooth muscle-type KATP channels with ABCC9. KCNJ11 forms the channel pore while ABCC9 is required for activation and regulation.
Specifications
| Phospho-Kir6.2 (Thr224) | |
| Polyclonal | |
| Unconjugated | |
| Kcnj11 | |
| AI842722; ATP-sensitive inward rectifier potassium channel 11; AW491124; beta-cell inward rectifier subunit; BIR; HHF2; IKATP; Inward rectifier K(+) channel Kir6.2; inwardly rectifying potassium channel KIR6.2; IRK11; KATP; KATP channel; KCNJ11; Kir6.2; mBIR; MGC133230; MODY13; PHHI; potassium channel inwardly rectifing subfamily J member 11; potassium channel, inwardly rectifying subfamily J member 11; potassium channel, inwardly rectifying subfamily J, member 11; potassium inwardly rectifying channel, subfamily J, member 11; potassium inwardly-rectifying channel subfamily J member 11; potassium inwardly-rectifying channel, subfamily J, member 11; potassium voltage-gated channel subfamily J member 11; TNDM3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16514, 3767 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14654, Q61743 | |
| Kcnj11 | |
| Peptide sequence around phosphorylation site of threonine 224 (K-T-TP-S-P) derived from Human Kir6.2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction