missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Kir3.1 (Ser185) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538572
This item is not returnable.
View return policy
Description
This potassium channel is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. This receptor plays a crucial role in regulating the heartbeat.
Specifications
| Phospho-Kir3.1 (Ser185) | |
| Polyclonal | |
| Unconjugated | |
| KCNJ3 | |
| KCNJ3, GIRK1, Kcnf3, KIR3.1, GIRK-1, HGIRK1, KGA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16519, 3760, 50599 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P48549, P63250, P63251 | |
| KCNJ3 | |
| A synthetic phosphopeptide derived from human GIRK1/KIR31/KCNJ3 around the phosphorylation site of Ser185 (K-M-SP-Q-P) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction