missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-JunD (Ser255) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104821
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of JunD only when phosphorylated at Serine 255.
JunD is a transcription factor which is coded by JunD gene. JunD transcriptional activity is modulated by phosphorylation in response to cellular stress via the c-Jun N terminal kinase (JNK)/Stress-Activated Protein Kinase (SAPK) family of protein kinase. Jun proteins are capable of forming dimers with Fos, ATF and CREB family transcription factors to form the AP-1 complex that differentially regulates a variety of target genes involved in cellular growth, proliferation, differentiation and apoptosis. Also, JunD plays an important role in metabolism via modulation of IGF-1 signaling pathway.
Specifications
| Phospho-JunD (Ser255) | |
| Polyclonal | |
| Unconjugated | |
| JUND | |
| activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jund; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 16478, 24518, 3727 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15066, P17535, P52909 | |
| JUND | |
| A synthesized peptide derived from human JUND(Accession P17535), corresponding to amino acid residues around phosphorylated Ser255. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction