missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-JunB (Ser259) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105511
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of JunB only when phosphorylated at Ser259.
JunB, a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5'-TGA[CG]TCA-3'. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.
Specifications
| Phospho-JunB (Ser259) | |
| Polyclonal | |
| Unconjugated | |
| Junb | |
| activator protein 1; AP-1; Jun B; jun B proto-oncogene; Junb; Jun-b; Jun-B oncogene; JunB proto-oncogene, AP-1 transcription factor subunit; myD21; Oncogene Jun-B; Transcription factor jun-B | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 16477, 24517, 3726 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09450, P17275, P24898 | |
| Junb | |
| A synthesized peptide derived from human JUNB(Accession P17275), corresponding to amino acid residues around phosphorylated Ser259. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction