missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-JUN (Ser73) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£109.00 - £249.00
Specifications
| Antigen | JUN (Ser73) |
|---|---|
| Concentration | 330 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235296
|
Proteintech
28891-1-AP-100UL |
100 μL |
£280.00 £249.00 / 100µL Save £31.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236607
|
Proteintech
28891-1-AP-20UL |
20 μL |
£125.00 £109.00 / 20µL Save £16.00 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-JUN (Ser73) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-JUN (Ser73). This antibody recognizes human, mouse, rat antigen. The Phospho-JUN (Ser73) antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| JUN (Ser73) | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| AP-1; AP1; c-Jun | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 330 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 3725 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title