missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Jak3 (Tyr980, Tyr981) Recombinant Rabbit Monoclonal Antibody (JAK3Y980981-E10), PE, Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£184.00 - £464.00

Specifications

Antigen Phospho-Jak3 (Tyr980, Tyr981)
Clone JAK3Y980981-E10
Applications Flow Cytometry
Classification Recombinant Monoclonal
Conjugate PE
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17157038
View Documents
Invitrogen™
MA537449
10 Tests
£184.00
Pack of 10
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17176788
GREENER_CHOICE
View Documents
Invitrogen™
MA537201
100 Tests
£464.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Janus kinase 3 (JAK3) phosphorylates members of the STAT (signal transduction and activators of transcription) family. JAK3 is predominantly expressed in immune cells and transduces a signal in response to activation of interleukin receptors. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. However, active JAK1 may be required for complex responses. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex.
Specifications

Specifications

Phospho-Jak3 (Tyr980, Tyr981)
Flow Cytometry
PE
Rabbit
Human
P52333
3718
A synthetic phospho-peptide corresponding to residues surrounding Tyr980/981 of human Jak3.
Primary
4°C
JAK3
JAK3Y980981-E10
Recombinant Monoclonal
Liquid
RUO
PBS with 0.20% BSA and 0.09% sodium azide
Breast-JAK3; fae; JAK3; JAK-3; JAK3_HUMAN; JAK3B; JAKL; Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; L JAK; Leukocyte janus kinase; LJAK; L-JAK; RATJAK3; Tyrosine-protein kinase JAK3; wil
JAK3
IgG κ
Protein A/G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.