missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Jak3 (Tyr980, Tyr981) Recombinant Rabbit Monoclonal Antibody (JAK3Y980981-E10), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA537203
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Janus kinase 3 (JAK3) phosphorylates members of the STAT (signal transduction and activators of transcription) family. JAK3 is predominantly expressed in immune cells and transduces a signal in response to activation of interleukin receptors. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. However, active JAK1 may be required for complex responses. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex.
Spezifikation
| Phospho-Jak3 (Tyr980, Tyr981) | |
| Recombinant Monoclonal | |
| APC | |
| JAK3 | |
| Breast-JAK3; fae; JAK3; JAK-3; JAK3_HUMAN; JAK3B; JAKL; Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; L JAK; Leukocyte janus kinase; LJAK; L-JAK; RATJAK3; Tyrosine-protein kinase JAK3; wil | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 3718 | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| JAK3Y980981-E10 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| P52333 | |
| JAK3 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr980/981 of human Jak3. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur