missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-JAK2/JAK3 (Tyr966, Tyr939) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5118652
This item is not returnable.
View return policy
Description
Phospho-JAK2/JAK3 (Tyr966, Tyr939) Polyclonal antibody specifically detects Phospho-JAK2/JAK3 (Tyr966, Tyr939) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
| Phospho-JAK2/JAK3 (Tyr966, Tyr939) | |
| Polyclonal | |
| Unconjugated | |
| JAK2 | |
| Breast-JAK3; fae; Fd17; JAK2; JAK-2; JAK3; JAK-3; JAK3_HUMAN; JAK3B; JAKL; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; JTK10; Leukocyte janus kinase; LJAK; L-JAK; RATJAK3; THCYT3; tyrosine-protein kinase JAK2; Tyrosine-protein kinase JAK3; wil | |
| A synthesized peptide derived from human JAK2/3 around the phosphorylation site of Tyr966/939. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60674, P52333, Q62120, Q62137, Q62689, Q63272 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16452, 16453, 24514, 25326, 3717, 3718 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction