Learn More
Invitrogen™ Phospho-IRS1 (Ser636) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA11055
Description
PA1-1055 detects phospho-IRS-1 (Ser636) in mouse and human cells. PA1-1055 has been successfully used in WB procedures. By Western blot, this antibody detects a ~170 kDa protein representing phospho-IRS-1 (Ser636) from 3T3 L1 cells treated with insulin for 10 minutes. The PA1-1055 immunogen is a synthetic phosphopeptide corrsponding to residues D(631) Y M P M (pS) P K S V S A P Q Q I(646) of human IRS-1. The peptide sequence is 100% conserved in rats, and mice. This peptide (Cat. # PEP-185) is available for use in neutralization and control experiments.
IRS-1, a major substrate of the insulin receptor, is phosphorylated in response to stimulation of cells by insulin, insulin-like growth factor 1 (IGF-1) and interleukin 4 (IL-4). IRS-1 is phosphorylated on serine, threonine and tyrosine residues in a variety of tissues. An insulin-sensitive serine/threonine kinase casein kinase II mediates a portion of the insulin-stimulated serine/threonine phosphorylation of overexpressed IRS-1 in vivo. Thr 502 is identified as the major casein kinase II-catalyzed phosphorylation site in rat IRS-1, and Ser 99 is an additional phosphorylation site catalyzed by casein kinase II. Thus, casein kinase II-catalyzed phosphorylation of IRS-1 may be a component of the intracellular insulin signaling cascade. IRS-1 contains three putative binding sites for 14-3-3 (Ser 270, Ser 374 and Ser 641) and the motif around Ser 270 is located in the phosphortyrosine binding domain of IRS-1, which is responsible for the interaction with the insulin receptor.
Specifications
| Phospho-IRS1 (Ser636) | |
| Polyclonal | |
| Unconjugated | |
| IRS1 | |
| G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16367, 3667 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1 mg/mL BSA and 0.05% sodium azide | |
| P35568, P35569 | |
| IRS1 | |
| Synthetic phosphopeptide corrsponding to residues D(631) Y M P M (pS) P K S V S A P Q Q I(646) of human IRS-1. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.