missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IRF7 (Ser477) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564834
Additional Details : Weight : 0.01000kg
Description
Phospho-IRF7 (Ser477) Polyclonal Antibody detects endogenous levels of IRF7 only when phosphorylated at Ser477.
Interferons (IFNs) are involved in a multitude of immune interactions during viral infections and play a major role in both the induction and regulation of innate and adaptive antiviral mechanisms. During infection, host-virus interactions signal downstream molecules such as transcription factors such as IFN regulatory factor-3 (IRF3) which can act to stimulate transcription of IFN-a/b genes. IRF7 has been shown to play a role in the transcriptional activation of virus-inducible cellular genes, including interferon beta chain genes. IRF7 play a major role in the innate immune pathway, interacting with the Toll-like receptor (TLR) adaptor proteins MyD88 and Tirp/TRAM and functioning as an intermediate TLR4 and TLR9 signaling. There are at least four differentially spliced isoforms of IRF7, although their function has not been clearly established.Specifications
Phospho-IRF7 (Ser477) | |
Polyclonal | |
Unconjugated | |
IRF7 | |
IMD39; Interferon regulatory factor 7; interferon regulatory factor-7H; Irf7; IRF-7; IRF7A; IRF7B; IRF7C; IRF7H; IRF-7H | |
Rabbit | |
Affinity chromatography | |
RUO | |
293624, 3665, 54123 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P70434, Q92985 | |
IRF7 | |
A synthesized peptide derived from human IRF-7 (Phospho-Ser477) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |