missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-IRF5 (Ser437) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564760
This item is not returnable.
View return policy
Description
Phospho-IRF5 (Ser437) Polyclonal Antibody detects endogenous levels of IRF5 only when phosphorylated at Ser437.
This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist.
Specifications
| Phospho-IRF5 (Ser437) | |
| Polyclonal | |
| Unconjugated | |
| IRF5 | |
| AW491843; Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF5; IRF-5; mirf5; OTTHUMP00000212907; SLEB10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27056, 296953, 3663 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| P56477, Q13568 | |
| IRF5 | |
| A synthesized peptide derived from human IRF5 (Phospho-Ser437). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction