missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-IRF3 (Ser396) Recombinant Rabbit Monoclonal Antibody (PSH06-75)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557852

Product Code. 30284467

  • £371.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: HT-29 transfected with 2.5μg/mL Poly (I:C) for 6 hours cell lysate, J774A.1 transfected with 2.5μg/mL Poly (I:C) for 6 hours cell lysate, human testis tissue, HT-29 cells transfected withPoly (I:C). Tissue Specificity: Low tissue specificity. Subcellular Location: Cytoplasm, Nucleus, Mitochondrion. Sequence Similarities: 83% Mouse/Rat. Predicted band size: 47 kDa.

IFR3 (interferon regulatory factor 3) is a key transcriptional regulator of type I interferon (IFN)-dependent immune responses. It plays a critical role in the innate immune response against DNA and RNA viruses. IFR3 regulates the transcription of type I IFN genes (IFN-alpha and IFN-beta) and IFN-stimulated genes (ISG) by binding to an interferon-stimulated response element (ISRE) in their promoters. It acts as a more potent activator of the IFN-beta gene than the IFN-alpha gene and plays a critical role in both the early and late phases of the IFNA/B gene induction. IFR3 is found in an inactive form in the cytoplasm of uninfected cells and following viral infection, double-stranded RNA, or toll-like receptor signaling. It is phosphorylated by IKBKE and TBK1 kinases. This induces a conformational change, leading to its dimerization and nuclear localization and association with CREB binding protein to form dsRNA-activated factor 1 (DRAF1), a complex which activates the transcription of the type I IFN and ISG genes. Mutations in the gene can result in encephalopathy, acute, infection-induced, herpes-specific 7.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-IRF3 (Ser396)
Recombinant Monoclonal
1.0 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P70671, Q14653
IRF3
Synthetic phospho-peptide corresponding to residues surrounding Ser396 of human IRF3.
100 μL
Primary
Human, Mouse
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
PSH06-75
Unconjugated
IRF3
C920001K05Rik; IIAE7; Interferon regulatory factor 3; IRF3; IRF-3; IRF3 (phospho S386); IRF3 (pS386); IRF3 (pSer386); IRF3 phospho; IRF3 phospho S386; IRF3 phospho Ser386; phospho IRF3
Rabbit
Protein A
RUO
3661, 54131
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.