missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IRF3 (Ser386) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537612
This item is not returnable.
View return policy
Description
A suggested positive control for Western blot is 293 or COLO cells.
Phospho-IRF3 (Ser386) Polyclonal antibody specifically detects Phospho-IRF3 (Ser386) in Human samples. It is validated for Western Blot.
Specifications
| Phospho-IRF3 (Ser386) | |
| Polyclonal | |
| Unconjugated | |
| IRF3 | |
| C920001K05Rik; IIAE7; Interferon regulatory factor 3; IRF3; IRF-3; IRF3 (phospho S386); IRF3 (pS386); IRF3 (pSer386); IRF3 phospho; IRF3 phospho S386; IRF3 phospho Ser386; phospho IRF3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3661 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14653 | |
| IRF3 | |
| Peptide sequence around phosphorylation site of Serine 386(A-S-S(p)-L-E) derived from Human IRF-3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction