missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-IRAK4 (Thr342) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105122
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of IRAK4 only when phosphorylated at Thr342.
IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. Overexpression of IRAK-4 activates NF-kappa-B and MAPK pathways. IRAK-4 interacts with and phosphorylates IRAK-1. IRAK-4-deficient animals are completely resistant to the challenge with LPS. Animals and humans lacking IRAK-4 are impaired in their responses to viral and bacterial challenges. Members in IRAK/Pelle family play a central role in IL-1R/TLR mediated inflammatory responses and in innate immunity.
Specifications
| Phospho-IRAK4 (Thr342) | |
| Polyclonal | |
| Unconjugated | |
| IRAK4 | |
| 8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 266632, 51135 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R4K2, Q9NWZ3 | |
| IRAK4 | |
| A synthesized peptide derived from human IRAK4(Accession Q9NWZ3), corresponding to amino acid residues around phosphorylated Thr342. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction