missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IP3 Receptor 1 (Ser1764) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539790
This item is not returnable.
View return policy
Description
Intracellular channel that mediates calcium release from the endoplasmic reticulum following stimulation by inositol 1,4,5-trisphosphate. Involved in the regulation of epithelial secretion of electrolytes and fluid through the interaction with AHCYL1 (By similarity). Plays a role in ER stress-induced apoptosis. Cytoplasmic calcium released from the ER triggers apoptosis by the activation of CaM kinase II, eventually leading to the activation of downstream apoptosis pathways.
Specifications
| Phospho-IP3 Receptor 1 (Ser1764) | |
| Polyclonal | |
| Unconjugated | |
| ITPR1 | |
| Inositol 1,4,5,-Triphosphate Receptor Type I, IP3R, IP3R1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16438, 25262, 3708 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14643, P11881, P29994 | |
| ITPR1 | |
| A synthetic phosphopeptide derived from human IP3R1 around the phosphorylation site of Ser1764 (R-E-SP-L-T) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction