missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-ILK (Thr173) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114587
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of ILK only when phosphorylated at Thr173.
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| Phospho-ILK (Thr173) | |
| Polyclonal | |
| Unconjugated | |
| ILK | |
| 59 kDa serine/threonine-protein kinase; AA511515; epididymis secretory protein Li 28; ESTM24; HEL-S-28; ILK; ILK1; ILK-1; ILK2; ILK-2; integrin binding protein kinase; Integrin linked ILK; integrin linked kinase; integrin-linked kinase; integrin-linked kinase-2; integrin-linked protein kinase; loc; lost-contact; main squeeze; msq; P59; p59ILK; zgc:65842; zilk | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 16202, 170922, 3611 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O55222, Q13418, Q99J82 | |
| ILK | |
| A synthesized peptide derived from human ILK around the phosphorylation site of Thr173. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction