missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-IKK beta (Tyr199) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538280
This item is not returnable.
View return policy
Description
Inhibitor of nuclear factor kappa-B kinase subunit beta (IKK beta) is a serine kinase that plays an essential role in the NF-kaapa-B signaling pathway which is activated by mulitple stimuli such as inflammatory cytokines, bacterial, or viral products, DNA damages, or other cellular stresses. IKK beta acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Mutations affecting the IKK beta gene can result in Immunodeficiency 15 (IMD15).
Specifications
| Phospho-IKK beta (Tyr199) | |
| Polyclonal | |
| Unconjugated | |
| IKBKB | |
| Inhibitor of nuclear factor kappa B kinase beta subunit, I-kappa-B-kinase beta, IkBKB, IKKB, I-kappa-B kinase 2, IKK2, Nuclear factor NF-kappa-B inhibitor kinase beta, NFKBIKB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16150, 3551, 84351 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14920, O88351, Q9QY78 | |
| IKBKB | |
| A synthetic phosphopeptide derived from human IKK-beta around the phosphorylation site of Tyr199 (Q-K-YP-T-V) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction