missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-IKK alpha/beta (Ser176, Ser180) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517593
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
IKK alpha/beta is encoded by CHUK (conserved helix-loop-helix ubiquitous kinase) and IKKB (inhibitor of kappa light polypeptide gene enhancer in B-cells) gene located in chromosome 10 and 8 respectively in humans. Upon stimulation by agents that activate the NF-kB pathway such as TNF alpha, the two subunits of IKK complex, IKK alpha and IKK -beta alongside a regulator NEMO (IKK gamma) phosphorylates IkB proteins. This targets the later for ubiquitin mediated degradation and phosphorylation of NF-kB leading to subsequent translocation to the nucleus. Phosphorylation at serine 176 and 180 located in the activation loop causes a confirmation change that is required for the activation of the kinases.
Specifications
| Phospho-IKK alpha/beta (Ser176, Ser180) | |
| Polyclonal | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O14920, O15111, O88351, Q60680, Q9QY78 | |
| CHUK, Ikbkb | |
| Phosphopeptide corresponding to a region surrounding pSer177/181 of IKKβ. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| CHUK | |
| AI132552; AI256658; AIM-1; CHUK; Chuk1; component of inhibitor of nuclear factor kappa B kinase complex; conserved helix-loop-helix ubiquitous kinase; EC 2.7.11.10; Fbx24; Fbxo24; I kappa B kinase 1; i kappa-B kinase alpha; IkappaB kinase; I-kappa-B kinase 1; I-kappa-B kinase 2; IkappaB kinase alpha; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; I-kappa-B-kinase beta; IkB kinase alpha subunit; IkB kinase-alpha; IkB kinase-beta; ikBKA; Ikbkb; IKK alpha; IKK[b]; IKK1; IKK2; IKK-2; IKKA; IKK-A; IKK-a kinase; IKK-alpha; Ikkb; IKK-B; IKKbeta; IKK-beta; IMD15; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta; inhibitor of kappaB kinase beta; inhibitor of nuclear factor kappa B kinase beta subunit; inhibitor of nuclear factor kappa B kinase subunit beta; inhibitor of nuclear factor kappa-B kinase subunit alpha; Inhibitor of nuclear factor kappa-B kinase subunit beta; NFKBIKA; NFKBIKB; Nuclear factor NFkappaB inhibitor kinase alpha; nuclear factor NF-kappa-B inhibitor kinase alpha; Nuclear factor NF-kappa-B inhibitor kinase beta; TCF16; TCF-16; TCF17; Transcription factor 16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1147, 12675, 16150, 309361, 3551, 84351 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction