missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HTRA2 (Ser212) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118695
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of HtrA2 only when phosphorylated at Ser212.
The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity (1-3). Recently, a serine protease has been isolated, which is released from the mitochondria upon induction of apoptosis by apoptotic stimuli. This protein called Omi/HtrA2 is a mitochondrial protein that binds to IAP (4). Like Smac/DIABLO, the mature Omi protein contains a conserved IAP-binding motif (AVPS) at its N terminus. The deregulated expression of Omi in the cytoplasm of mammalian cells induces apoptosis indicating that Omi could participate in the mitochondrial apoptotic pathway.
Specifications
| Phospho-HTRA2 (Ser212) | |
| Polyclonal | |
| Unconjugated | |
| HTRA2 | |
| AI481710; High temperature requirement protein A2; HtrA serine peptidase 2; HtrA2; HtrA-like serine protease; mnd2; motor neuron degeneration 2; OMI; Omi stress-regulated endoprotease; PARK13; protease, serine, 25; Prss25; Serine protease 25; serine protease HTRA2, mitochondrial; serine protease OMI; serine proteinase OMI | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 27429, 297376, 64704 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43464, Q9JIY5 | |
| HTRA2 | |
| A synthesized peptide derived from human HTRA2(Accession O43464), corresponding to amino acid residues around phosphorylated Ser212. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction