Learn More
Invitrogen™ Phospho-Histone H2A.X (Ser139) Recombinant Mouse Monoclonal Antibody (CR55T33), Invitrogen™
Mouse Recombinant Monoclonal Antibody
£135.00 - £360.00
Specifications
| Antigen | Phospho-Histone H2A.X (Ser139) |
|---|---|
| Clone | CR55T33 |
| Concentration | 1.0 mg/mL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30131125
|
Invitrogen™
740078M |
100 μL |
£360.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30131659
|
Invitrogen™
740078M20UG |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. Applications Tested: This CR55T33 clone has been tested for flow cytometry at 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Phospho-Histone H2A.X (Ser139) Recombinant Monoclonal antibody specifically detects Phospho-Histone H2A.X (Ser139) in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Phospho-Histone H2A.X (Ser139) | |
| 1.0 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| AW228881; gamma H2AX; gammaH2ax; H2A histone family member X; H2A histone family, member X; H2A.X; H2A.X variant histone; H2A/X; h2afx; H2ax; H2AX histone; Hist5-2ax; histone 5 protein 2ax; histone H2A.x; Histone H2a/x; Histone H2AX; RGD1566119; similar to H2A histone family, member X; zgc:56329 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| CR55T33 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P16104, P27661 | |
| 15270, 3014 | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| H2AX |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.