missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HIPK1/HIPK2/HIPK3 (Tyr352, Tyr361, Tyr359) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564584
This item is not returnable.
View return policy
Description
Phospho-HIPK1/HIPK2/HIPK3 (Tyr352, Tyr361, Tyr359) Polyclonal Antibody detects endogenous levels of HIPK1/HIPK2/HIPK3 only when phosphorylated at Tyr352, Tyr361, Tyr359.
The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms.
Specifications
| Phospho-HIPK1/HIPK2/HIPK3 (Tyr352, Tyr361, Tyr359) | |
| Polyclonal | |
| Unconjugated | |
| HIPK1 | |
| 1110014O20Rik; 1110062K04Rik; androgen receptor-interacting nuclear protein kinase; ANPK; B230339E18Rik; DYRK6; EMBL:ABO47653.1}; fas-interacting serine/threonine-protein kinase; FIST; FIST3; HCMV-interacting protein; hHIPk2; Hipk1; hipk1 {ECO:0000312; HIPK2; HIPK3; homeodomain interacting protein kinase 1; homeodomain interacting protein kinase 1-like protein; homeodomain interacting protein kinase 2; homeodomain interacting protein kinase 3; homeodomain-interacting protein kinase 1; Homeodomain-interacting protein kinase 2; homeodomain-interacting protein kinase 3; Homolog of protein kinase YAK1; KIAA0630; mir-1902; Myak; Nbak1; Nbak2; nbak3; nuclear body associated kinase 2b; nuclear body-associated kinase 1; Nuclear body-associated kinase 2; nuclear body-associated kinase 3; nuclear serine/threonine protein kinase; PKY; PRO0593; protein kinase Myak; Sialophorin tail-associated nuclear serine/threonine-protein kinase; Stank; YAK1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10114, 15257, 15258, 15259, 204851, 28996, 362342, 365895, 83617 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A4L9P5, O88850, O88904, Q86Z02, Q9ERH7, Q9H2X6, Q9H422, Q9QZR5 | |
| HIPK1, HIPK2, HIPK3 | |
| A synthesized peptide derived from human HIPK1/2/3 (Phospho-Tyr352/361/359). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction