missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HGS (Tyr216) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540200
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
HGS is involved in intracellular signal transduction mediated by cytokines and growth factors. When associated with STAM, it suppresses DNA signaling upon stimulation by IL-2 and GM-CSF. It could be a direct effector of PI3-kinase in vesicular pathway via early endosomes and may regulate trafficking to early and late endosomes by recruiting clathrin. HGS may concentrate ubiquitinated receptors within clathrin-coated regions. It is involved in down-regulation of receptor tyrosine kinase via multivesicular body (MVBs) when complexed with STAM. This complex binds ubiquitin and acts as sorting machinery that recognizes ubiquitinated receptors and transfers them to further sequential lysosomal sorting/trafficking processes. HGS may contribute to the efficient recruitment of SMADs to the activin receptor complex.
Specifications
| Phospho-HGS (Tyr216) | |
| Polyclonal | |
| Unconjugated | |
| HGS | |
| hepatocyte growth factor-regulated tyrosine kinase substrate; HGF-regulated tyrosine kinase substrate; HGNC:4897; Hgr; Hgs; Hrs; Hrs2; human growth factor-regulated tyrosine kinase substrate; protein pp110; SNAP-25-interacting protein Hrs-2; ZFYVE8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15239, 56084, 9146 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| O14964, Q99LI8, Q9JJ50 | |
| HGS | |
| Synthesized peptide derived from human Hrs around the phosphorylation site of Y216. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction