missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HEC1 (Ser165) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534978
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, Blocking peptide (GTX70013-PEP). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
Specifications
| Phospho-HEC1 (Ser165) | |
| Polyclonal | |
| Unconjugated | |
| NDC80 | |
| 2610020P18Rik; AV139730; HEC; HEC1; HEC-1; HEC2; HEC4; Highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; HsHec1; hsNDC80; kinetochore associated 2; kinetochore protein Hec1; Kinetochore protein NDC80 homolog; kinetochore-associated protein 2; KNTC2; NDC80; NDC80 homolog, kinetochore complex component; NDC80 homolog, kinetochore complex component (S. cerevisiae); NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; NDC80, kinetochore complex component; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; TID3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10403, 301701, 67052 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.127 mg/mL | |
| PBS with no preservative | |
| O14777, Q9D0F1 | |
| NDC80 | |
| Synthetic phosphopeptide corresponding to the amino acids around Ser 165 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rodent, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction