missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HDAC6 (Ser22) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105035
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of HDAC6 only when phosphorylated at Serine 22.
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Plays a central role in microtubule-dependent cell motility via deacetylation of tubulin.
Specifications
| Phospho-HDAC6 (Ser22) | |
| Polyclonal | |
| Unconjugated | |
| HDAC6 | |
| CPBHM; FLJ16239; Hd6; HDA6; Hdac5; Hdac6; histone deacetylase 5; histone deacetylase 6; Histone deacetylase mHDA2; JM21; KIAA0901; mHDA2; PPP1R90; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Sfc6 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 10013, 15185 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UBN7, Q9Z2V5 | |
| HDAC6 | |
| A synthesized peptide derived from human HDAC6(Accession Q9UBN7), corresponding to amino acid residues around phosphorylated Ser22. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction