missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HDAC4/HDAC5/HDAC7 (Ser246, Ser259, Ser155) Monoclonal Antibody (B.544.8)

Description
It is not recommended to aliquot this antibody.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to mouse HDAC7 gene whose protein promotes repression mediated via the transcriptional corepressor SMRT. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | Phospho-HDAC4/HDAC5/HDAC7 (Ser246, Ser259, Ser155) |
| Applications | ChIP Assay, Western Blot |
| Classification | Monoclonal |
| Clone | B.544.8 |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 |
| Gene | HDAC4 |
| Gene Accession No. | P56524, Q6NZM9, Q8C2B3, Q8WUI4, Q9UQL6, Q9Z2V6 |
| Gene Alias | 4932408F19Rik; 5830434K02Rik; AHO3; AI047285; AI426555; Antigen NY-CO-9; BDMR; HA6116; HD4; HD5; HD7; HD7a; Hdac4; HDAC-4; HDAC5; Hdac7; hdac-7; HDAC7A; HDACA; HDAC-A; Histone deacetylase 4; histone deacetylase 5; Histone deacetylase 7; histone deacetylase 7A; histone deacetylase A; histone deacetylase mHDA1; KIAA0288; KIAA0600; mFLJ00062; mHDA1; mKIAA0600; NY-CO-9 |
| Gene Symbols | HDAC4, HDAC5, HDAC7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?