missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HCLS1 (Tyr397) Recombinant Rabbit Monoclonal Antibody (HS1Y397-F12), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537162
This item is not returnable.
View return policy
Description
Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. May also be involved in the regulation of gene expression. [UniProt].
Specifications
| Phospho-HCLS1 (Tyr397) | |
| Recombinant Monoclonal | |
| APC | |
| HCLS1 | |
| AW213261; CTTNL; HCLS1; hematopoietic cell specific Lyn substrate 1; hematopoietic cell-specific Lyn substrate 1; Hematopoietic lineage cell-specific protein; HS1; LckBP1; p75 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 15163, 3059 | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| HS1Y397-F12 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| P14317, P49710 | |
| HCLS1 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr397 of human phospho HS1. | |
| 100 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction