missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HBP1 (Ser402) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599136
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of HBP1 only when phosphorylated at Sersine 402.
The HMG-box protein-1 (HBP1) is a member of the HMG family of transcription factors, which are characterized by the presence of a conserved protein motif, the high mobility group (HMG) 1 box, that mediates DNA binding. HBP1 binds to the tumor suppressor proteins Rb and p130 and initiates cell cycle arrest. Terminal cell differentiation requires this initial cell cycle arrest followed by the coordinated expression of genes defined as tissue-specific markers. Along with initiating the commitment to cell differentiation, the continued activity of HBP1 abrogates the expression of tissue-specific genes by associating with the MyoD proteins. In muscle cell differentiation, the MyoD family of transcription factors, which include Myf5, MyoD and myogenein, induce the expression of these cell-type specific proteins and contribute to the development of cell phenotypes. The progression of terminal differentiation is, therefore, dependent on both a decrease in HBP1 activity and the corresponding activation of MyoD-induced gene transcription.
Specifications
| Phospho-HBP1 (Ser402) | |
| Polyclonal | |
| Unconjugated | |
| HBP1 | |
| 1700058O05Rik; BE629963; C330012F01Rik; C86454; Hbp1; High mobility group box transcription factor 1; high mobility group protein 1; HMG box transcription factor 1; HMG box-containing protein 1; Hmgb1; HMG-box containing protein 1; HMG-box transcription factor 1 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 26959 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60381 | |
| HBP1 | |
| A synthesized peptide derived from human HBP1(Accession O60381), corresponding to amino acid residues around phosphorylated Ser402. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction