missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GYS1 (Ser641) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121310
This item is not returnable.
View return policy
Description
Positive test controls include: 293T, NIH/3T3, C6. Immunogen sequence: PASVP.
Glycogen Synthase 1 (GYS1) catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Phospho-GYS1 (Ser641) | |
| Polyclonal | |
| Unconjugated | |
| Gys1 | |
| EC 2.4.1.11; glycogen [starch] synthase, muscle; glycogen synthase; glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 1, muscle; glycogen synthase 3, brain; GSY; GYS; GYS1; Gys3; MGS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14936, 2997, 690987 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.28 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| A2RRU1, P13807, Q9Z1E4 | |
| Gys1 | |
| A synthetic phosphorylated peptide around S641 of human Glycogen synthase 1 (GYS1) (NP_0020942). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction