missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-GSK3B (Tyr216) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£125.00 - £280.00
Specifications
| Antigen | Phospho-GSK3B (Tyr216) |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17402373
|
Proteintech
29125-1-AP-20UL |
20 μL |
£125.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17492363
|
Proteintech
29125-1-AP-100UL |
100 μL |
£280.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-GSK3B (Tyr216) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-GSK3B (Tyr216). This antibody recognizes human antigen. The Phospho-GSK3B (Tyr216) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-GSK3B (Tyr216) | |
| Polyclonal | |
| Rabbit | |
| 2932 | |
| Primary |
| Western Blot, ELISA | |
| Unconjugated | |
| Human | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title