missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GRK2 (Tyr86) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106089
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of GRK2 only when phosphorylated at Tyr86.
ADRBK1 (GRK2) is a serine/threonine kinase. Many of the G protein-coupled receptors are known to be phosphorylated by G protein-dependent receptor kinases (GRKs). G protein-coupled receptor kinases (GRKs) are important regulators of G protein-coupled receptors(GPCRs). GRK2 (83 kDa), one of six members of this family that have been identified, is ubiquitously expressed in mammals. After binding to their ligand and interacting with heterotrimeric G proteins, GPCRs (e.g., a2-adrenergic receptor) are phosphorylated by GRKs. Internalization of the GPCRs, regulated by beta-arrestin-1, leads to activation of the Ras - Raf - ERK1&2 signaling pathway. GRK2 activity is tightly controlled by different mechanisms including phosphorylation by kinases such as PKC, Src and ERK1&2, as well as interaction with various proteins. ERK phosphorylates and thus inactivates GRK2 on serine 670 in a negative feedback mechanism.
Specifications
| Phospho-GRK2 (Tyr86) | |
| Polyclonal | |
| Unconjugated | |
| GRK2 | |
| ADRBK1; Adrbk-1; adrenergic beta receptor kinase 1; adrenergic receptor kinase, beta 1; adrenergic, beta, receptor kinase 1; BARK; BARK1; Bark-1; beta ARK; Beta-adrenergic receptor kinase 1; beta-adrenergic receptor kinase 1 beta ARK1; beta-AR kinase-1; betaARK1; BETA-ARK1; beta-ARK-1; EC 2.7.1.126; G protein-coupled receptor kinase 2; G-protein coupled receptor kinase 2; G-protein-coupled receptor kinase 2; grk 2; Grk2; GRK-2 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 110355, 156, 25238 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P25098, P26817, Q99MK8 | |
| GRK2 | |
| A synthesized peptide derived from human GRK2(Accession P25098), corresponding to amino acid residues around phosphorylated Tyr86. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction