missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-GNAZ (Ser16) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5118619
This item is not returnable.
View return policy
Description
Phospho-GNAZ (Ser16) Polyclonal antibody specifically detects Phospho-GNAZ (Ser16) in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Phospho-GNAZ (Ser16) | |
| Polyclonal | |
| Unconjugated | |
| GNAZ | |
| AI847979; G protein subunit alpha z; g(x) alpha chain; GNAZ; guanine nucleotide binding protein (G protein), alpha z polypeptide; Guanine nucleotide binding protein alpha; guanine nucleotide binding protein, alpha z polypeptide; guanine nucleotide binding protein, alpha z subunit; guanine nucleotide-binding protein G(z) subunit alpha; GXA; Gz; Gz-alpha; transducin alpha | |
| A synthesized peptide derived from human Gz-alpha around the phosphorylation site of Ser16. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70443, P19086, P19627 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14687, 25740, 2781 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction