missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GluR2 (Ser880) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595671
This item is not returnable.
View return policy
Description
Antigen Affinity Purified from Pooled Serum.
The ion channels activated by glutamate are typically divided into two classes. Those that are sensitive to N-methyl-D-aspartate (NMDA) are designated NMDA receptors (NMDAR) while those activated by alpha-amino-3-hydroxy-5 -methyl-4-isoxalone propionic acid (AMPA) are known as AMPA receptors (AMPAR). The AMPAR are comprised of four distinct Glutamate Receptor Subunits designated (GluR1-4) and they play key roles in virtually all excitatory neurotransmission in the brain. The GluR2 subunit is thought to play a key role in forms of synaptic plasticity such as LTD.
Specifications
| Phospho-GluR2 (Ser880) | |
| Polyclonal | |
| Unconjugated | |
| Gria2 | |
| AMPA selective glutamate receptor; AMPA-selective glutamate receptor 2; GluA2; Glur2; GluR-2; GLURB; GluR-B; GluR-K2; glutamate ionotropic receptor AMPA type subunit 2; glutamate receptor; glutamate receptor 2; glutamate receptor B; glutamate receptor B flip isoform; glutamate receptor ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA2 (alpha 2); GRIA2; HBGR2; OTTHUMP00000165324; OTTHUMP00000220942; OTTHUMP00000220943 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29627 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| ~0.25 mg/mL | |
| 10mM HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P19491 | |
| Gria2 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser880 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction