Learn More
Invitrogen™ Phospho-GluR1 (Ser845) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ OPA104118
Description
This antibody is predicted to react with human, mouse and non-human primate based on 100% sequence homology. In Western blot, this antibody detects a ~100 kDa protein representing phospho-GluR1 (pSer845) in rat brain lysate. Immunolabeling is blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide.
GluR1 (glutamate receptor 1) is an ionotropic glutamate receptor. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter, L-glutamate, induces a conformation change that leads to the opening of the cation channel and converts the chemical signal to an electrical impulse. Then, the receptor desensitizes rapidly and enters a transiet inactive state characterized by the presence of a bound agonist. In the presence of CACNG4, CACNG7, or CACNG8 GluR1 shows resensitization which is characterized by a delayed accumulation of current flux upon continued application of glutamate.
Specifications
| Phospho-GluR1 (Ser845) | |
| Polyclonal | |
| Unconjugated | |
| GRIA1 | |
| 2900051M01Rik; AI853806; AMPA 1; AMPA glutamate receptor; AMPA receptor GluR1/A; AMPA-selective glutamate receptor 1; Glr1; Glr-1; GluA1; GLUH 1; GLUH1; GluR K1; GLUR1; Glur-1; GluR1 flip; GluR1 flop; GLURA; GluR-A; GluRK1; gluR-K1; glutamate ionotropic receptor AMPA type subunit 1; glutamate receptor; glutamate receptor 1; glutamate receptor A; glutamate receptor ionotropic, AMPA 1; glutamate receptor subunit GluR1; glutamate receptor, ionotropic, AMPA 1; glutamate receptor, ionotropic, AMPA1 (alpha 1); GPRC1A; Gria 1; Gria1; GRM1A; HBGR1; HGBR1; HIPA; HIPA1; MGC13325; MGC133252; NMDA1; NMDAR1; NR1; OTTHUMP00000160643; OTTHUMP00000165781; OTTHUMP00000224241; OTTHUMP00000224242; OTTHUMP00000224243; RP11-350O14.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14799, 50592 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| 0.25 mg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P19490, P23818 | |
| GRIA1 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser845 of rat GRIA1 conjugated to KLH. | |
| 150 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.