missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GIT2 (Tyr592) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105211
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of GIT2 only when phosphorylated at Tyr592.
GTPase-activating protein for the ADP ribosylation factor family.
Specifications
| Phospho-GIT2 (Tyr592) | |
| Polyclonal | |
| Unconjugated | |
| GIT2 | |
| 1500036H07Rik; 5830420E16Rik; 6430510B20Rik; 9630056M03Rik; ARF GAP GIT2; ARF GTPase activating protein 2; ARF GTPase-activating protein GIT2; B230104M05Rik; CAT2; CAT-2; cool associated tyrosine phosphorylated-2; cool-associated, tyrosine phosphorylated protein 2; cool-interacting tyrosine-phosphorylated protein 2; G protein-coupled receptor kinase interacting ArfGAP 2; G protein-coupled receptor kinase-interactor 2; GIT ArfGAP 2; Git2; GRK-interacting protein 2; KIAA0148; mKIAA0148; Paxillin kinase linker; PKL | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 9815 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14161 | |
| GIT2 | |
| A synthesized peptide derived from human GIT2(Accession Q14161), corresponding to amino acid residues around phosphorylated Tyr592. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction