missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GCN2 (Thr899) Recombinant Rabbit Monoclonal Antibody (JE00-51)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556574
This item is not returnable.
View return policy
Description
Phospho-GCN2 (Thr899) Recombinant Monoclonal Antibody for Western Blot
EIF2AK4 (GCN2) is a metabolic-stress sensing protein kinase that phosphorylates the alpha subunit of eukaryotic translation initiation factor-2 in response to low amino acid availability. It plays a role as an activator of the integrated stress response required for adapatation to amino acid starvation. Mutations in the gene lead to pulmonary vennocclusive disease 2.
Specifications
| Phospho-GCN2 (Thr899) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| D4A7V9, Q9P2K8 | |
| EIF2AK4 | |
| Synthetic phosphopeptide corresponding to residues surrounding Thr899 of Human GCN2. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| JE00-51 | |
| Unconjugated | |
| EIF2AK4 | |
| 2610011M03; EIF2AK4; eIF-2-alpha kinase GCN2; eukaryotic translation initiation factor 2 alpha kinase 4; eukaryotic translation initiation factor 2-alpha kinase 4; Gcn2; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2; Kiaa1338; mGCN2; PVOD2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 114859, 440275 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction