missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-GATA2 (Ser401) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15574521
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15574521 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574521 Supplier Invitrogen™ Supplier No. PA537583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control for Western blot is JK cells.

GATA2 (GATA binding protein 2) belongs to the GATA family of transcription factors. GATA proteins play a very important role in signaling pathways controlling proliferation and differentiation of erythroid progenitor cells. There are 6 transcription factors in the GATA family: GATA1 to GATA6. GATA2 is a transcriptional activator regulating endothelin-1 gene expression in endothelial cells. It also appears to be involved in differentiation of other tissue types (uro-genital and nervous) during embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-GATA2 (Ser401)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GATA2
Gene Accession No. O09100, P23769
Gene Alias DCML; EMBL:AAK51128.1}; Endothelial transcription factor GATA-2; FLJ45948; GATA binding protein 2; GATA binding protein 3; GATA2; Gata-2; gata2 {ECO:0000312; GATA3; GATA-binding factor 3; GATA-binding protein 2; GATA-bindning protein 2; HDR; HDRS; IMD21; MGC2306; MONOMAC; NFE1B; trans-acting T-cell-specific transcription factor GATA-3; uterine GATA-4 transcription factor
Gene Symbols GATA2
Host Species Rabbit
Immunogen Peptide sequence around phosphorylation site of Serine 401(K-M-S(p)-N-K) derived from Human GATA2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14461, 2624
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.