missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-GAP43 (Ser41) Polyclonal Antibody

Phospho-GAP43 (Ser41) Polyclonal Antibody
Brand: Thermo Scientific PA5114577
This item is not returnable.
View return policy
Description
Phospho-GAP43 (Ser41) Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Phospho-GAP43 (Ser41) | |
| Polyclonal | |
| Unconjugated | |
| GAP43 | |
| Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 14432, 2596, 29423 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P06837, P07936, P17677 | |
| GAP43 | |
| A synthesized peptide derived from human GAP43 around the phosphorylation site of Ser41. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction