missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GABRG2 (Ser327) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14739
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, chicken, mouse, human and non-human primates based on 100% sequence homology. In Western blot, a band at ~45 kDa is detected. Immunolabeling can be blocked by lambda-phosphatase treatment.
This gene encodes a gamma-aminobutyric acid (GABA) receptor. GABA is the major inhibitory neurotransmitter in the mammlian brain, where it acts at GABA-A receptors, which are ligand-gated chloride channels. GABA-A receptors are pentameric, consisting of proteins from several subunit classes: alpha, beta, gamma, delta and rho. Mutations in this gene have been associated with epilepsy and febrile seizures. Multiple transcript variants encoding different isoforms have been identified for this gene.
Specifications
| Phospho-GABRG2 (Ser327) | |
| Polyclonal | |
| Unconjugated | |
| Gabrg2 | |
| CAE2; ECA2; GABA(A) receptor subunit gamma-2; GABA(A) receptor, gamma 2; GABAA-R; GABRA2; Gabrg2; Gabrg-2; gamma2; gamma-aminobutyrate receptor type A gamma-2 subuni; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 2; gamma-aminobutyric acid A receptor gamma 2; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid receptor subunit gamma-2; gamma-aminobutyric acid type A receptor gamma2 subunit; gamma-aminobutyric acid type A receptor subunit gamma2; GEFSP3; LOC553402 protein; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit gamma-2; si:ch211-145n14.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14406, 29709 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P18508, P22723 | |
| Gabrg2 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser327 of rat GABRG2 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction