missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GABRB3 (Ser408, Ser409) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535390
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, chicken, human, mouse, non-human primate, Xenopus and zebrafish based on 100% sequence homology. This antibody contains enough material to conduct 10 mini-Western blots.
Gamma-amino butyric acid (GAMA) is a strong inhibitor of neuron transmission and is found throughout the central nervous system. GABA is formed by the decarboxylation of glutamate, resulting in the formation of one of two distinct types; GABA-A or GABA-B.The binding of GABA to GABA-A receptors results in the increase transportation of chlorine ions into the neuron. This influx results in signal transmission repression. Studies have shown that if cell to cell signaling goes unchecked due to a lack of GABA, a system overload can resulting in blackouts, rapid body contractions, seizures and epilepsy.
Specifications
| Phospho-GABRB3 (Ser408, Ser409) | |
| Polyclonal | |
| Unconjugated | |
| GABRB3 | |
| A230092K12Rik; AW049585; beta3; cleft palate 1; Cp1; ECA5; GABA(A) receptor subunit beta-3; GABAA receptor beta-3 subunit; GABA-alpha receptor beta-2 subunit; GABR B3; Gabrb3; Gabrb-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 3; gamma-aminobutyric acid (GABA) A receptor, subunit beta 3; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 3; gamma-aminobutyric acid A receptor beta 3; Gamma-aminobutyric acid receptor beta 3; gamma-aminobutyric acid receptor subunit beta-3; gamma-aminobutyric acid receptor, subunit beta 3; gamma-aminobutyric acid type A receptor beta3 subunit; gamma-aminobutyric acid type A receptor subunit beta3; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit beta-3; MGC9051; testis gamma-aminobutyric acid receptor subunit beta 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14402, 24922 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| ∼0.25 mg/mL | |
| 0.01M HEPES with 0.1mg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P63079, P63080 | |
| GABRB3 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser408/409 of rat GABRB3 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction