missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-GABRB1 (Ser434) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538068
Code : NEW
Additional Details : Weight : 0.00900kg
Description
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel.Specifications
Phospho-GABRB1 (Ser434) | |
Polyclonal | |
Unconjugated | |
GABRB1 | |
GABA(A) receptor, GABA(A)R beta-1, GBRB1, Gamma-aminobutyric-acid receptor beta-1 subunit precursor, GABA-RB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14400, 25450, 2560 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P18505, P50571, P15431 | |
GABRB1 | |
A synthetic phosphopeptide derived from human GABA-RB around the phosphorylation site of Ser434 (R-A-SP-Q-L) | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |