missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-GAB2 (Tyr614) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105940
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Gab2 only when phosphorylated at Tyr614.
This gene is a member of the GRB2-associated binding protein (GAB) gene family. These proteins contain pleckstrin homology (PH) domain, and bind SHP2 tyrosine phosphatase and GRB2 adapter protein. They act as adapters for transmitting various signals in response to stimuli through cytokine and growth factor receptors, and T- and B-cell antigen receptors. The protein encoded by this gene is the principal activator of phosphatidylinositol-3 kinase in response to activation of the high affinity IgE receptor. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
Specifications
| Phospho-GAB2 (Tyr614) | |
| Polyclonal | |
| Unconjugated | |
| GAB2 | |
| AI463667; D130058I17Rik; GAB2; Grb2 associated binder 2; GRB2 associated binding protein 2; GRB2-associated binder 2; GRB2-associated binding protein 2; GRB2-associated-binding protein 2; GRB3-3; Growth factor receptor bound protein 2-associated protein 2; KIAA0571; p97; PH domain-containing adaptor molecule p97; pp100 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 14389, 84477, 9846 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9EQH1, Q9UQC2, Q9Z1S8 | |
| GAB2 | |
| A synthesized peptide derived from human GAB2(Accession Q9UQC2), corresponding to amino acid residues around phosphorylated Tyr614. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction