missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Fumarase (Thr90) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106222
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of FH only when phosphorylated at T90.
The protein encoded by this gene is an enzymatic component of the tricarboxylic acid cycle, or Krebs cycle, and catalyzes the formation of L-malate from fumarate. It exists in both a cytosolic form and an N-terminal extended form, differing only in the translation start site used. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension generates the same form as in the cytoplasm. It is similar to some thermostable class II fumarases and functions as a homotetramer. Mutations in this gene can cause fumarase deficiency and lead to progressive encephalopathy.
Specifications
| Phospho-Fumarase (Thr90) | |
| Polyclonal | |
| Unconjugated | |
| FH | |
| EF-3; Fh; Fh1; Fh-1; FMRD; fumarase; fumarate hydratase; fumarate hydratase 1; fumarate hydratase, mitochondrial; HLRCC; LRCC; MCL; MCUL1 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 14193, 14194, 2271, 24368 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P07954, P14408, P97807 | |
| FH, Fh1 | |
| A synthesized peptide derived from human FH(Accession P07954), corresponding to amino acid residues around phosphorylated Thr90. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction