Learn More
Invitrogen™ Phospho-FOXA2 (Thr156) Recombinant Superclonal™ Antibody (10HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 710680
Description
This antibody is predicted to react with mouse, bovine and zebrafish based on sequence homology. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
FOXA2 is a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific genes such as albumin and transthyretin. They also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. FOXA2 has been linked to sporadic cases of maturity-onset diabetes of the young. Transcript variants encoding different isoforms have been identified.
Specifications
| Phospho-FOXA2 (Thr156) | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q9Y261 | |
| FOXA2 | |
| Synthetic peptide corresponding to human FOXA2, aa 153-160, phosphorylated at Threonine 156. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 10HCLC | |
| Unconjugated | |
| FOXA2 | |
| FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3B; transcription factor; transcription factor 3B; transcription factor Foxa2; WHN | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3170 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.