missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Filamin A (Tyr1604) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105527
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Filamin A only when phosphorylated at Tyr1604.
Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.
Specifications
| Phospho-Filamin A (Tyr1604) | |
| Polyclonal | |
| Unconjugated | |
| Flna | |
| 450 kDa protein; ABP-280; ABPX; actin binding protein 280; actin-binding protein 280; alpha filamin; alpha-filamin; CSBS; CVD1; dilated pupils 2; Dilp2; Endothelial actin-binding protein; Endothelial actin-binding protein 280); F730004A14Rik; filamin; filamin 1; filamin A; filamin A, alpha; filamin, alpha; Filamin1; filamin-1; FilaminA; Filamin-A; FLN; FLN1; Flna; FLN-A; FMD; GENA 379; gizzard filamin; MNS; NHBP; Non-muscle filamin; OPD; OPD1; OPD2; RGD1560614; XLVD; XMVD | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 192176, 2316, 293860 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P21333, Q8BTM8 | |
| Flna | |
| A synthesized peptide derived from human FLNA(Accession P21333), corresponding to amino acid residues around phosphorylated Tyr1604. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction